4-Chlor-2-hydroxybenzonitril 98%
Lieferant: Apollo Scientific
|
Achtung
|
OR46050-1G
OR46050-5G
OR46050-10G
OR46050-25G
APOSOR46050-1GEA
36.4
EUR
APOSOR46050-1G
APOSOR46050-5G
APOSOR46050-10G
APOSOR46050-25G
4-Chlor-2-hydroxybenzonitril 98%
4-Chlor-2-hydroxybenzonitril
Formel:
C₇H₄ClNO Molecular Weight: 153,5700 g/mol Schmelzpunkt: 155 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234255 CAS-Nummer: 30818-28-1 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...