4-Methyl-1H-1,2,3-triazole
Lieferant: Apollo Scientific
|
Achtung
|
OR46054-1G
OR46054-250MG
APOSOR46054-1GEA
424
EUR
APOSOR46054-1G
APOSOR46054-250MG
4-Methyl-1H-1,2,3-triazole
4-Methyl-1H-1,2,3-triazole
Formel:
C₃H₅N₃ Molecular Weight: 83,0919 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD14706836 CAS-Nummer: 27808-16-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...