2-Chlor-3,4-pyridindiamin
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3,4-Diamino-2-chloropyridine, 2-Chloropyridine-3,4-diamine
OR46057-25G
APOSOR46057-25GEA
125
EUR
APOSOR46057-25G
2-Chlor-3,4-pyridindiamin
2-Chloropyridine-3,4-diamine
Formel:
C₅H₆ClN₃ Molecular Weight: 143,5742 g/mol Schmelzpunkt: 158…163 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07363739 CAS-Nummer: 39217-08-8 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...