Methyl-3-chloro-4-cyanobenzoate 98%
Lieferant: Apollo Scientific
Synonyme:
2-CHLORO-4-(METHOXYCARBONYL)BENZONITRILE, Methyl 3-chloro-4-cyanobenzoate, 3-CHLORO-4-CYANO-BENZOIC ACID METHYL ESTER
OR46071-100MG
OR46071-250MG
OR46071-1G
APOSOR46071-100MGEA
84.4
EUR
APOSOR46071-100MG
APOSOR46071-250MG
APOSOR46071-1G
Methyl-3-chloro-4-cyanobenzoate 98%
Methyl-3-chloro-4-cyanobenzoate
Formel:
C₉H₆ClNO₂ Molecular Weight: 195,602 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12756074 CAS-Nummer: 214759-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...