5-Bromo-2,3-dimethylpyridine

Lieferant: Apollo Scientific
Gefahr

OR46088-1G OR46088-250MG
APOSOR46088-1GEA 21.9 EUR
APOSOR46088-1G APOSOR46088-250MG
5-Bromo-2,3-dimethylpyridine
5-Bromo-2,3-dimethylpyridine
Formel: C₇H₈BrN
Molecular Weight: 186,0491 g/mol
Siedepunkt: 40 °C/0,3mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD11656270
CAS-Nummer: 27063-90-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR