3-Brom-5-iodanisol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
1-Bromo-3-iodo-5-methoxybenzene
OR46092-1G
OR46092-5G
APOSOR46092-1GEA
134
EUR
APOSOR46092-1G
APOSOR46092-5G
3-Brom-5-iodanisol
3-Brom-5-iodanisol
Formel:
C₇H₆BrIO Molecular Weight: 312,930 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD12547766 CAS-Nummer: 915412-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...