3-Brom-4-chlor-5-nitropyridin
Lieferant: Apollo Scientific
|
Achtung
|
OR46093-100G
OR46093-25G
OR46093-5G
APOSOR46093-100GEA
865
EUR
APOSOR46093-100G
APOSOR46093-25G
APOSOR46093-5G
3-Brom-4-chlor-5-nitropyridin
3-Bromo-4-chloro-5-nitropyridine
Formel:
C₅H₂BrClN₂O₂ Schmelzpunkt: 49…50 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07368836 CAS-Nummer: 31872-63-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...