4-Chlor-3-methoxyphenylboronsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(4-Chloro-3-methoxyphenyl)boronic acid
OR46099-1G
OR46099-25G
OR46099-5G
APOSOR46099-1GEA
21.9
EUR
APOSOR46099-1G
APOSOR46099-25G
APOSOR46099-5G
4-Chlor-3-methoxyphenylboronsäure 98%
4-Chlor-3-methoxyphenylboronsäure
Formel:
C₇H₈BClO₃ Molecular Weight: 186,4000 g/mol Schmelzpunkt: 232…234 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01318965 CAS-Nummer: 89694-47-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...