4-Formyl-2-methoxypyridin 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Methoxypyridine-4-carbaldehyde, 2-Methoxyisonicotinaldehyde

OR46110-1G OR46110-5G
APOSOR46110-1GEA 46.4 EUR
APOSOR46110-1G APOSOR46110-5G
4-Formyl-2-methoxypyridin 98%
4-Formyl-2-methoxypyridin
Formel: C₇H₇NO₂
Molecular Weight: 137.14 g/mol
Schmelzpunkt: 36 °C
Lagertemperatur: Kühlschrank
MDL: MFCD06410680
CAS-Nummer: 72716-87-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR