5-Methoxynicotinaldehyde 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methoxy-pyridinecarboxaldehyde
OR46113-1G
OR46113-25G
OR46113-5G
APOSOR46113-1GEA
63.8
EUR
APOSOR46113-1G
APOSOR46113-25G
APOSOR46113-5G
5-Methoxynicotinaldehyde 97%
5-Methoxynicotinaldehyde
Formel:
C7H7NO2 Molecular Weight: 137,1400 g/mol Schmelzpunkt: 33…34 °C Flammpunkt: 100 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD07437946 CAS-Nummer: 113118-83-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...