7-Bromo-1H-imidazo[4,5-c]pyridine 95%
Lieferant: Apollo Scientific
|
Gefahr
|
OR46120-1G
OR46120-250MG
APOSOR46120-1GEA
120
EUR
APOSOR46120-1G
APOSOR46120-250MG
7-Bromo-1H-imidazo[4,5-c]pyridine 95%
7-Bromo-1H-imidazo[4,5-c]pyridine
Formel:
C₆H₄BrN₃ Molecular Weight: 198.0200 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09834585 CAS-Nummer: 90993-26-3 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...