4-Brom-2-methyl-6-nitroanilin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-5-brom-3-nitrotoluol, 4-Brom-6-nitro-o-toluidin

OR4613-5G OR4613-1G OR4613-25G
APOSOR4613-5GEA 159 EUR
APOSOR4613-5G APOSOR4613-1G APOSOR4613-25G
4-Brom-2-methyl-6-nitroanilin
4-Brom-2-methyl-6-nitroanilin
Formel: C₇H₇BrN₂O₂
Molecular Weight: 231.05 g/mol
Schmelzpunkt: 143 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052919
CAS-Nummer: 77811-44-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR