ω,3-Dibromacetophenon 95%
Lieferant: Apollo Scientific
Synonyme:
2,3'-Dibromacetophenon, 2-Brom-1-(3-bromphenyl)-1-ethanon, 3-Bromphenacylbromid, α,3-Dibromacetophenon
OR46152-25G
OR46152-1G
OR46152-5G
APOSOR46152-25GEA
0
EUR
APOSOR46152-25G
APOSOR46152-1G
APOSOR46152-5G
ω,3-Dibromacetophenon 95%
ω,3-Dibromacetophenon
Formel:
C₈H₆Br₂O Molecular Weight: 277.94 g/mol Schmelzpunkt: 51 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082698 CAS-Nummer: 18523-22-3 UN: 3261 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...