4-(Methylsulfonyl)benzylbromid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
α-Brom-4-(methylsulfonyl)toluol
OR46201-5G
OR46201-100G
OR46201-25G
OR46201-1G
APOSOR46201-5GEA
79.9
EUR
APOSOR46201-5G
APOSOR46201-100G
APOSOR46201-25G
APOSOR46201-1G
4-(Methylsulfonyl)benzylbromid 97%
4-(Methylsulfonyl)benzylbromid
Formel:
C₈H₉BrO₂S Molecular Weight: 249.13 Schmelzpunkt: 82…86 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00185849 CAS-Nummer: 53606-06-7 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...