6-Bromo-4-methylpyridin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-6-bromo-4-methylpyridine
OR46205-1G
OR46205-5G
OR46205-10G
APOSOR46205-1GEA
21.9
EUR
APOSOR46205-1G
APOSOR46205-5G
APOSOR46205-10G
6-Bromo-4-methylpyridin-2-amine
6-Bromo-4-methylpyridin-2-amine
Formel:
C₆H₇BrN₂ Molecular Weight: 187.05 g/mol Schmelzpunkt: 115…116 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD11036206 CAS-Nummer: 73895-98-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...