4-Amino-3-brom-2-chlorpyridin

Lieferant: Apollo Scientific

Synonyme: 3-Brom-2-chlor-4-pyridylamin

OR46206-25G OR46206-5G
APOSOR46206-25GEA 473 EUR
APOSOR46206-25G APOSOR46206-5G
4-Amino-3-brom-2-chlorpyridin
4-Amino-3-bromo-2-chloropyridine
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
Schmelzpunkt: 148…153 °C
Lagertemperatur: Kühlschrank
MDL: MFCD09263478
CAS-Nummer: 215364-85-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR