3H-Imidazo[4,5-c]pyridine 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Aza-1H-benzoimidazole, 5-Azabenzimidazole
OR46225-1G
OR46225-5G
OR46225-25G
APOSOR46225-1GEA
29
EUR
APOSOR46225-1G
APOSOR46225-5G
APOSOR46225-25G
3H-Imidazo[4,5-c]pyridine 96%
3H-Imidazo[4,5-c]pyridine
Formel:
C₆H₅N₃ Molecular Weight: 119,13 g/mol Schmelzpunkt: 168…173 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051808 CAS-Nummer: 272-97-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...