1H-Pyrazol-3-carbaldehyd 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Formylpyrazole, Pyrazole-3-carboxaldehyde, 3-Formyl-1H-pyrazole
OR46247-100G
OR46247-25G
OR46247-5G
OR46247-1G
APOSOR46247-100GEA
224
EUR
APOSOR46247-100G
APOSOR46247-25G
APOSOR46247-5G
APOSOR46247-1G
1H-Pyrazol-3-carbaldehyd 97%
1H-Pyrazol-3-carbaldehyd
Formel:
C₄H₄N₂O Molecular Weight: 96,09 g/mol Schmelzpunkt: 149…150 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129925 CAS-Nummer: 3920-50-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...