2-Amino-5-bromthiophenol

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-Amino-5-bromobenzenethiol

OR46252-1G OR46252-250MG
APOSOR46252-1GEA 0 EUR
APOSOR46252-1G APOSOR46252-250MG
2-Amino-5-bromthiophenol
2-Amino-5-bromthiophenol
Formel: C₆H₆BrNS
Molecular Weight: 204.800 g/mol
Schmelzpunkt: 113…115 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00839467
CAS-Nummer: 23451-95-8
UN: 1759
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR