(-)-Corey lactone benzoate 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: (3aR,4S,5R,6aS)-4-(Hydroxymethyl)-2-oxohexahydro-2H-cyclopenta[b]furan-5-yl Benzoate, (3aR,4S,5R,6aS)-5-(Benzoyloxy)hexahydro-4-(hydroxymethyl)-2H-cyclopenta[b]furan-2-one

OR46255-1G OR46255-5G
APOSOR46255-1GEA 79.9 EUR
APOSOR46255-1G APOSOR46255-5G
(-)-Corey lactone benzoate 98%
(-)-Corey lactone benzoate
Formel: C₁₅H₁₆O₅
Molecular Weight: 276.29 g/mol
Schmelzpunkt: 120 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00674102
CAS-Nummer: 39746-00-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR