3-Brombenzhydrazid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Bromobenzoic hydrazide

OR4644-100G
APOSOR4644-100GEA 194 EUR
APOSOR4644-100G
3-Brombenzhydrazid 98%
3-Brombenzhydrazid
Formel: C₇H₇BrN₂O
Molecular Weight: 215,05 g/mol
Schmelzpunkt: 156…158 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007600
CAS-Nummer: 39115-96-3
EINECS: 254-298-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR