4-(Trimethylsilyloxy)benzaldehyd 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-[(Trimethylsilyl)oxy]benzaldehyde

OR4648-25G
APOSOR4648-25GEA 85.7 EUR
APOSOR4648-25G
4-(Trimethylsilyloxy)benzaldehyd 97%
4-(Trimethylsilyloxy)benzaldehyd
Formel: C₁₀H₁₄O₂Si
Molecular Weight: 194,3025 g/mol
Siedepunkt: 83…84 °C/3mmHg
Lagertemperatur: Kühlschrank
MDL: MFCD00040723
CAS-Nummer: 1012-12-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR