3-Methoxypyridin-4-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Amino-3-methoxypyridine
OR46502-1G
OR46502-100MG
OR46502-250MG
OR46502-500MG
APOSOR46502-1GEA
0
EUR
APOSOR46502-1G
APOSOR46502-100MG
APOSOR46502-250MG
APOSOR46502-500MG
3-Methoxypyridin-4-amine
3-Methoxypyridin-4-amine
Formel:
C6H8N2O Lagertemperatur: Raumtemperatur |
MDL:
MFCD09991938 CAS-Nummer: 52334-90-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...