Methyl-2-chlor-6-methoxyisonicotinat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Methyl 2-chloro-6-methoxypyridine-4-carboxylate, Methyl 2-chloro-6-methoxyisonicotinate
OR46530-1G
OR46530-5G
OR46530-25G
APOSOR46530-1GEA
23.2
EUR
APOSOR46530-1G
APOSOR46530-5G
APOSOR46530-25G
Methyl-2-chlor-6-methoxyisonicotinat
Methyl-2-chlor-6-methoxyisonicotinat
Formel:
C₈H₈ClNO₃ Molecular Weight: 201,61 g/mol Schmelzpunkt: 97,5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173929 CAS-Nummer: 42521-10-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...