6-Iod-o-toluylsäure 95%

Lieferant: Apollo Scientific
Achtung

OR46539-1G OR46539-250MG OR46539-5G
APOSOR46539-1GEA 122 EUR
APOSOR46539-1G APOSOR46539-250MG APOSOR46539-5G
6-Iod-o-toluylsäure 95%
6-Iod-o-toluylsäure
Formel: C₈H₇IO₂
Molecular Weight: 262,05 g/mol
Schmelzpunkt: 112…116 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01310823
CAS-Nummer: 54811-50-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR