Hexaketocyclohexane octahydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: Cyclohexanehexone octahydrate, Triquinoyl octahydrate, Cyclohexane-1,2,3,4,5,6-hexone octahydrate

OR46546-10G OR46546-2G
APOSOR46546-10GEA 280 EUR
APOSOR46546-10G APOSOR46546-2G
Hexaketocyclohexane octahydrate
Hexaketocyclohexane octahydrate
Formel: C₆O₆·8H₂O
Molecular Weight: 312,18 g/mol
Schmelzpunkt: 99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00149074
CAS-Nummer: 7255-28-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR