2-Methyl-2-propanyl-(2-bromethyl)carbamat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(BOC-amino)ethyl bromide

OR46552-1G OR46552-25G OR46552-5G
APOSOR46552-1GEA 29.2 EUR
APOSOR46552-1G APOSOR46552-25G APOSOR46552-5G
2-Methyl-2-propanyl-(2-bromethyl)carbamat 97%
tert-Butyl (2-bromoethyl)carbamate
Formel: C₇H₁₄BrNO₂
Molecular Weight: 224,10 g/mol
Schmelzpunkt: 30…32 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD02683428
CAS-Nummer: 39684-80-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR