4-Chloro-3-nitrophenyl methyl sulphone

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Chloro-4-(methylsulphonyl)-2-nitrobenzene

OR46580-10G OR46580-1G OR46580-5G
APOSOR46580-10GEA 68.4 EUR
APOSOR46580-10G APOSOR46580-1G APOSOR46580-5G
4-Chloro-3-nitrophenyl methyl sulphone
4-Chloro-3-nitrophenyl methyl sulphone
Formel: C₇H₆ClNO₄S
Molecular Weight: 235.64 g/mol
Schmelzpunkt: 125 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039754
CAS-Nummer: 97-07-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR