4-(Isobutyro)thioanisol

Lieferant: Apollo Scientific
Achtung

Synonyme: P-(methylthio)isobutyrophenone, 2-Methyl-1-[4-(methylsulphanyl)phenyl]propan-1-one

OR4659-100G OR4659-25G
APOSOR4659-100GEA 279 EUR
APOSOR4659-100G APOSOR4659-25G
4-(Isobutyro)thioanisol
4-(Isobutyro)thioanisol
Formel: C₁₁H₁₄OS
Molecular Weight: 194,2933 g/mol
Siedepunkt: 122…124 °C/2mmHg
Schmelzpunkt: 32…33 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094661
CAS-Nummer: 53207-58-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR