Bis(2-aminophenyl)sulphide
Lieferant: Apollo Scientific
|
Achtung
|
OR4660-25G
OR4660-5G
APOSOR4660-25GEA
0
EUR
APOSOR4660-25G
APOSOR4660-5G
Bis(2-aminophenyl)sulphide
Bis(2-aminophenyl)sulphide
Formel:
C₁₂H₁₂N₂S Schmelzpunkt: 85…88 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094662 CAS-Nummer: 5873-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...