2-Oxoglutarsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Ketoglutarsäure, α-Ketoglutarsäure

OR4662-100G OR4662-25G OR4662-500G
APOSOR4662-100GEA 125 EUR
APOSOR4662-100G APOSOR4662-25G APOSOR4662-500G
2-Oxoglutarsäure
2-Oxoglutarsäure
Formel: HOOCCH₂CH₂COCOOH
Molecular Weight: 146,1 g/mol
Schmelzpunkt: 113…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00004165
CAS-Nummer: 328-50-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR