N2-Cyclopentyl-2,3-pyridinediamine

Lieferant: Apollo Scientific

OR46624-1G OR46624-250MG OR46624-500MG
APOSOR46624-1GEA 451 EUR
APOSOR46624-1G APOSOR46624-250MG APOSOR46624-500MG
N2-Cyclopentyl-2,3-pyridinediamine
N2-Cyclopentyl-2,3-pyridinediamine
Formel: C₁₀H₁₅N₃
Molecular Weight: 177,25 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08692158
CAS-Nummer: 951523-75-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR