5-Chloro-1H-benzimidazol-2-amine
Lieferant: Apollo Scientific
OR46629-1G
OR46629-50MG
OR46629-250MG
OR46629-500MG
APOSOR46629-1GEA
519
EUR
APOSOR46629-1G
APOSOR46629-50MG
APOSOR46629-250MG
APOSOR46629-500MG
5-Chloro-1H-benzimidazol-2-amine
5-Chloro-1H-benzimidazol-2-amine
Formel:
C₇H₆ClN₃ Molecular Weight: 167,6 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00749332 CAS-Nummer: 5418-93-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...