6-Chlor-3,5-dibromanthranilsäure 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Amino-6-chloro-3,5-dibromobenzoic acid
OR4663-1G
OR4663-5G
APOSOR4663-1GEA
69.6
EUR
APOSOR4663-1G
APOSOR4663-5G
6-Chlor-3,5-dibromanthranilsäure 98%
6-Chlor-3,5-dibromanthranilsäure
Formel:
C₇H₄Br₂ClNO₂ Molecular Weight: 329.37 g/mol Schmelzpunkt: 212…215 °C Lagertemperatur: Raumtemperatur |
CAS-Nummer:
143769-25-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...