5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde

Lieferant: Apollo Scientific

OR46635-1G OR46635-250MG OR46635-500MG
APOSOR46635-1GEA 393 EUR
APOSOR46635-1G APOSOR46635-250MG APOSOR46635-500MG
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde
5-Chloro-1-methyl-3-phenyl-1H-pyrazole-4-carbaldehyde
Formel: C₁₁H₉ClN₂O
Molecular Weight: 220.66 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00244899
CAS-Nummer: 883-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR