Bis(3-hydroxyphenyl)disulphide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,3'-Dihydroxydiphenyl Disulphide, 3,3'-Dithiodiphenol, 3,3-Disulphanediyldiphenol
OR4665-5G
OR4665-25G
APOSOR4665-5GEA
163
EUR
APOSOR4665-5G
APOSOR4665-25G
Bis(3-hydroxyphenyl)disulphide
Bis(3-hydroxyphenyl)disulphide
Formel:
C₁₂H₁₀O₂S₂ Schmelzpunkt: 89…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03094663 CAS-Nummer: 21101-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...