3-(4-Chlorophenyl)oxirane-2-carboxylic acid

Lieferant: Apollo Scientific

OR46650-1G OR46650-100MG OR46650-250MG OR46650-500MG OR46650-5G
APOSOR46650-1GEA 980 EUR
APOSOR46650-1G APOSOR46650-100MG APOSOR46650-250MG APOSOR46650-500MG APOSOR46650-5G
3-(4-Chlorophenyl)oxirane-2-carboxylic acid
3-(4-Chlorophenyl)oxirane-2-carboxylic acid
Formel: C₉H₇ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD18089198
CAS-Nummer: 114380-31-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR