3-Bromo-5-chloro-2-hydroxybenzoic acid

Lieferant: Apollo Scientific

OR46653-1G OR46653-250MG OR46653-500MG
APOSOR46653-1GEA 432 EUR
APOSOR46653-1G APOSOR46653-250MG APOSOR46653-500MG
3-Bromo-5-chloro-2-hydroxybenzoic acid
3-Bromo-5-chloro-2-hydroxybenzoic acid
Formel: C₇H₄BrClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00985201
CAS-Nummer: 4068-58-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR