1-(4-Chlorophenyl)pyrrolidine-3-carboxylic acid 95%

Lieferant: Apollo Scientific

OR46661-1G OR46661-250MG OR46661-500MG
APOSOR46661-1GEA 693 EUR
APOSOR46661-1G APOSOR46661-250MG APOSOR46661-500MG
1-(4-Chlorophenyl)pyrrolidine-3-carboxylic acid 95%
1-(4-Chlorophenyl)pyrrolidine-3-carboxylic acid
Formel: C₁₁H₁₂ClNO₂
Lagertemperatur: Kühlschrank
MDL: MFCD09864922
CAS-Nummer: 933719-76-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR