Amino(phenyl)acetyl chloride

Lieferant: Apollo Scientific

OR46671-1G OR46671-100MG OR46671-250MG OR46671-500MG OR46671-2.5G OR46671-5G
APOSOR46671-1GEA 328 EUR
APOSOR46671-1G APOSOR46671-100MG APOSOR46671-250MG APOSOR46671-500MG APOSOR46671-2.5G APOSOR46671-5G
Amino(phenyl)acetyl chloride
Amino(phenyl)acetyl chloride
Formel: C₈H₈ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD03658916
CAS-Nummer: 39478-47-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR