2-Iodo-4-methoxybenzoyl chloride
Lieferant: Apollo Scientific
OR46676-250MG
OR46676-100MG
APOSOR46676-250MGEA
275
EUR
APOSOR46676-250MG
APOSOR46676-100MG
2-Iodo-4-methoxybenzoyl chloride
2-Iodo-4-methoxybenzoyl chloride
Formel:
C₈H₆ClIO₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD18905193 CAS-Nummer: 1201221-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...