5-Oxo-2-pyrrolidinecarbonyl chloride

Lieferant: Apollo Scientific

OR46689-1G OR46689-250MG OR46689-500MG
APOSOR46689-1GEA 693 EUR
APOSOR46689-1G APOSOR46689-250MG APOSOR46689-500MG
5-Oxo-2-pyrrolidinecarbonyl chloride
5-Oxo-2-pyrrolidinecarbonyl chloride
Formel: C₅H₆ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11938896
CAS-Nummer: 55478-53-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR