(2R,6S)-4-(3-Chloropropyl)-2,6-dimethylmorpholine

Lieferant: Apollo Scientific

OR46699-1G OR46699-250MG OR46699-500MG OR46699-2.5G
APOSOR46699-1GEA 458 EUR
APOSOR46699-1G APOSOR46699-250MG APOSOR46699-500MG APOSOR46699-2.5G
(2R,6S)-4-(3-Chloropropyl)-2,6-dimethylmorpholine
(2R,6S)-4-(3-Chloropropyl)-2,6-dimethylmorpholine
Formel: C₉H₁₈ClNO
Molecular Weight: 191.7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15731988
CAS-Nummer: 1130769-62-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR