3-Methyl-2-(1H-pyrrol-1-yl)phenol
Lieferant: Apollo Scientific
OR46704-250MG
OR46704-500MG
APOSOR46704-250MGEA
249
EUR
APOSOR46704-250MG
APOSOR46704-500MG
3-Methyl-2-(1H-pyrrol-1-yl)phenol
3-Methyl-2-(1H-pyrrol-1-yl)phenol
Formel:
C₁₁H₁₁NO Lagertemperatur: Raumtemperatur |
MDL:
MFCD21333135 CAS-Nummer: 1346497-81-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...