6,7-Dihydro-3H-cyclopenta[d]pyrimidin-4(5H)-one

Lieferant: Apollo Scientific

OR46721-1G OR46721-100MG OR46721-250MG OR46721-500MG OR46721-2.5G
APOSOR46721-1GEA 288 EUR
APOSOR46721-1G APOSOR46721-100MG APOSOR46721-250MG APOSOR46721-500MG APOSOR46721-2.5G
6,7-Dihydro-3H-cyclopenta[d]pyrimidin-4(5H)-one
6,7-Dihydro-3H-cyclopenta[d]pyrimidin-4(5H)-one
Formel: C₇H₈N₂O
Molecular Weight: 136,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00223593
CAS-Nummer: 5661-01-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR