3-(Hydroxymethyl)-1,3-benzothiazol-2(3H)-one

Lieferant: Apollo Scientific

OR46738-1G OR46738-250MG OR46738-500MG OR46738-5G
APOSOR46738-1GEA 137 EUR
APOSOR46738-1G APOSOR46738-250MG APOSOR46738-500MG APOSOR46738-5G
3-(Hydroxymethyl)-1,3-benzothiazol-2(3H)-one
3-(Hydroxymethyl)-1,3-benzothiazol-2(3H)-one
Formel: C₈H₇NO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD01074863
CAS-Nummer: 72679-97-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR