2,5-Dimethylphenylisocyanat 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,5-Xylylisocyanat

OR46742-1G OR46742-5G OR46742-25G
APOSOR46742-1GEA 21.9 EUR
APOSOR46742-1G APOSOR46742-5G APOSOR46742-25G
2,5-Dimethylphenylisocyanat 95%
2,5-Dimethylphenylisocyanat

  • May form small amount of sediment.

Formel: C₉H₉NO
Molecular Weight: 147,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00013854
CAS-Nummer: 40397-98-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR