Tetraoctylammoniumbromid

Lieferant: Apollo Scientific
Achtung

Synonyme: Tetra-n-octylammonium bromide

OR4675-100G OR4675-25G OR4675-500G
APOSOR4675-100GEA 26.1 EUR
APOSOR4675-100G APOSOR4675-25G APOSOR4675-500G
Tetraoctylammoniumbromid
Tetraoctylammoniumbromid
Formel: C₃₂H₆₈BrN
Molecular Weight: 546,8 g/mol
Schmelzpunkt: 95…99 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00011863
CAS-Nummer: 14866-33-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR