Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate

Lieferant: Apollo Scientific
Achtung

OR46756-1G OR46756-250MG
APOSOR46756-1GEA 157 EUR
APOSOR46756-1G APOSOR46756-250MG
Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate
Methyl-3-amino-6-cyclopropylthieno[2,3-b]pyridine-2-carboxylate
Formel: C₁₂H₁₂N₂O₂S
Molecular Weight: 248,31 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11841051
CAS-Nummer: 1135283-79-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR