Methyl-3,5-dibrom-4-methylbenzoat

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 3,5-dibromo-4-methylbenzoate

OR4685-100G OR4685-25G
APOSOR4685-100GEA 163 EUR
APOSOR4685-100G APOSOR4685-25G
Methyl-3,5-dibrom-4-methylbenzoat
Methyl-3,5-dibrom-4-methylbenzoat
Formel: C₉H₈Br₂O₂
Molecular Weight: 307,98 g/mol
Schmelzpunkt: 86…90 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010729
CAS-Nummer: 74896-66-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR